Adapters, bends
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.008.007
WITG1.008.007EA 28.1 EUR
WITG1.008.007
Adapters, bends
Onderdelen voor destillatie Destillatieadapters
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 105°. With 2 standard ground cones.