Splash heads, bent, acc. to Stutzer
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.425.005
WITG1.425.005EA 86.3 EUR
WITG1.425.005
Splash heads, bent, acc. to Stutzer
Onderdelen voor destillatie Destillatiekoppen
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two standard ground cones, inclined at an angle of 75°. Placed directly on the distillation flask to intercept droplets.