Funnels, with standard ground joint cone
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.650.014 1.650.029 1.650.129 1.650.130 1.650.150 1.650.019
WITG1.650.014EA 9.8 EUR
WITG1.650.014 WITG1.650.029 WITG1.650.129 WITG1.650.130 WITG1.650.150 WITG1.650.019
Funnels, with standard ground joint cone
Trechters Filtertrechters Trechters met conische slijpstukken
Funnels, made of boroslicate glass, with standard ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.