Funnels, with standard ground joint cone
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.651.019 1.651.029
WITG1.651.019EA 66.1 EUR
WITG1.651.019 WITG1.651.029
Funnels, with standard ground joint cone
Trechters Filtertrechters Trechters met conische slijpstukken
Funnels, made of boroslicate glass, with standard ground joint cone, one side flat.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.
Order Now
Specificaties
Environmentally Preferable