Drying tubes, bend
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.660.024 1.660.019
WITG1.660.024EA 25 EUR
WITG1.660.024 WITG1.660.019
Drying tubes, bend
Buisjes Droogbuizen
Borosilicate glass 3.3. Angled at 75° with ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.