Dropping funnels, cylindrical, with pressure balance
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.895.001 1.895.022 1.895.023 1.895.002 1.895.021
WITG1.895.001EA 80.8 EUR
WITG1.895.001 WITG1.895.022 WITG1.895.023 WITG1.895.002 WITG1.895.021
Dropping funnels, cylindrical, with pressure balance
Trechters Druppeltrechters
Dropping funnels, cylindrical, made of borosilicate glass, stopcock with glass key. With drip nozzle.
  • Graduated
  • Very good chemical resistance
  • Very good temperature resistance
With screw-thread retaling nut.