Gas burette, Bunte, Class A
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2.970.000
WITG2.970.000EA 281 EUR
WITG2.970.000
Gas burette, Bunte, Class A
Buretten
Gas burette made of borosilicate glass, accuracy Class A.
  • With resistant DIFFICO graduation
  • Main point ring graduation
ST-stopcock with double bore and screw-thread retaining nut.