Bulb pipettes, one mark, class AS
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3.535.125 3.535.040 3.535.100.BL 3.535.200.BL 3.535.110 3.535.130 3.535.140 3.535.075 3.535.038 3.535.800 3.535.035 3.535.160 3.535.700 3.535.600 3.535.750 3.535.170 3.535.055
WITG3.535.125EA 91.8 EUR
WITG3.535.125 WITG3.535.040 WITG3.535.100.BL WITG3.535.200.BL WITG3.535.110 WITG3.535.130 WITG3.535.140 WITG3.535.075 WITG3.535.038 WITG3.535.800 WITG3.535.035 WITG3.535.160 WITG3.535.700 WITG3.535.600 WITG3.535.750 WITG3.535.170 WITG3.535.055
Bulb pipettes, one mark, class AS
Pipetten Volpipetten
These bulb pipettes are made of soda lime glass.
  • With amber or blue stain DIFFICO ring marks and inscriptions
  • With tempered tips
  • With ISO-COLOR-Code, acc. to DIN (ISO 648)
  • Waiting time 15 s
  • Calibrated to contain "EX"
  • Conformity certified
Certificaten: DIN 12691