Culture flasks with rim, BIOGEN
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5.507.204 5.507.201 5.507.203 5.507.205
WITG5.507.204EA 46.8 EUR
WITG5.507.204 WITG5.507.201 WITG5.507.203 WITG5.507.205
Culture flasks with rim, BIOGEN
Kolven Cultuurflessen
Culture flask, made of borosilicate glass, Erlenmeyer form, designed for orbital shakers.
  • Neck with rim
  • Triple baffled shake flask
  • Excellent chemical and temperature resistance
Order Now
Specificaties
Environmentally Preferable