Evaporating dishes with spout
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5.530.010.B
WITG5.530.010.BEA 9.4 EUR
WITG5.530.010.B
Evaporating dishes with spout
Reservoirs/Schalen/Potten Uitdampschalen
Evaporating dishes are made of borosilicate glass, with spout.
  • Excellent chemical resistance
  • High temperature resistance up to +500 °C
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Certificaten: DIN 12336.