Aspirator bottles with screw caps and stopcock
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5.611.001 5.611.002 5.611.005 5.611.010
WITG5.611.001EA 139 EUR
WITG5.611.001 WITG5.611.002 WITG5.611.005 WITG5.611.010
Aspirator bottles with screw caps and stopcock
Flessen Doseerflessen
Borosilicate glass, clear, narrow neck.
  • With screw cap GL 45
  • Screw thread tubus GL 32 and valve tap
  • Good chemical und temperature resistance
Certificaten: DIN 12037