Woulff bottles
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5.810.500 5.810.010 5.810.005 5.810.002
WITG5.810.500EA 120 EUR
WITG5.810.500 WITG5.810.010 WITG5.810.005 WITG5.810.002
Woulff bottles
Flessen Woulff-flessen
Made of borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion gives relatively high resistance to temperature changes
With three necks.