Potassium chloride 3 mol/L 3 N aqueous solution

Leverancier: WTW
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassiumchloride

109706
WTWA109706EA 10.8 EUR
WTWA109706
Potassium chloride 3 mol/L 3 N aqueous solution
Potassium chloride
Formule: KCl
MW: 74.55 g/mol
Kookpunt: 1420
Smeltpunt: 773
Dichtheid: 1.13 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR