Spoon spatula, stainless steel
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5386-0022 5386-0025
BURK5386-0022EA 4.8 EUR
BURK5386-0022 BURK5386-0025
Spoon spatula, stainless steel
Spatels
The spatula-spoon can be used in many different applications. The double-sided multi-purpose spoon is a spoon on one side and a spatula on the other. This makes it possible to sample and process a variety of media such as powders, granulates, pastes and liquids. The spoon side of the spoon-spatula holds 4 ml.
  • Stainless steel AISI 304 (1.4301)
  • High-gloss polished
  • Double-sided function
Manufactured from a single piece of stainless steel, the spoon-spatula has no edges, crevices or undercuts. Because of this, product residue or cleaning agents cannot beMore Product Information