Masterflex® Fitting, Natural Polypropylene, Elbow, Hose Barb Union, 3/4" ID; 5/PK
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX40610-27
Your Organization's Price:
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/4" ID
Create 90° bends in flexible tubing—without the kinks!