Masterflex® Transfer Tubing, Platinum-Cured Silicone, 1/2" ID x 3/4" OD; 25 Ft
Leverancier: Avantor Fluid Handling
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
No need to worry about the heat - use for applications up to 460 °F.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- Tubing size: 1/2" ID x 3/4" OD
Applications: Pharmaceutical, biotechnology, aerospace, food and beverage, and general lab applications. <br />Characteristics: Platinum-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent. <br />Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI; exceeds 3A Sanitary Standards. Tubing is manufactured according to GMP. <br />Temperature range: -58 to 446 °F (-50 to 230 °C)<br />Sterilize: Autoclave or gamma irradiation.