Masterflex® Handheld Remote Controller, DB9(M) Connector; 6 ft (1.8 m) Cable
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX07528-80
Your Organization's Price:
Easily control your Masterflex® L/S® pumps while filling and dispensing.
  • Suitable for use with Masterflex® L/S® pumps
  • Long 10ft cable makes remote control simple
  • Connection Type: DB9(M)
These ergonomic handheld controllers are designed to control basic pump drive operations including start/stop, direction, and prime — depending on available drive functions. Route tubing through the handle for convenience in filling and dispensing applications. These handheld controllers accept Masterflex tubing sizes L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18, L/S 15, and L/S 24 (up to 7/16" nominal OD).
<b>Leveringsinformatie: </b>Includes 6-ft (1.8 m) cable with noted terminal connector.