Heavy-Duty Tubing Pinch Clamp, Polysulfone
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX20001-00
Your Organization's Price:
Durable design handles higher working pressures.
  • Quick installation with positive swing-away locking screw
  • Smooth contours allow for easy cleaning
Use this clamp with tubing or hose - it will meet the working pressure of either. High-strength polysulfone closure provides excellent durability, can be continually reused, and resists most chemicals. Simply align the hose within the clamp; mating surfaces on the clamp indicate complete closure. Typical applications include sterile filling, media bag connections, pumping, liquid control and high-purity fluid transfers. FDA-compliant polysulfone material can be sterilized by autoclave or gamma radiation.