Masterflex® Fitting, Polycarbonate, Three-Way, Large-Bore Stopcock with Male Luer Lock; 10/PK
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX30600-23
Your Organization's Price:
Verbeter de vloeistofstroom met een grotere doorlaat dan standaard luer-stopkranen.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Three-way flow pattern
Adapters met een grote diameter zijn perfect voor het verwerken van grotere stroomvolumes tijdens het proces, terwijl er verbindingen worden gebruikt die vergelijkbaar zijn met luer-systemen.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.