Masterflex® Fitting, Polypropylene, Straight, Female Luer to Hosebarb Adapter, 3/16" ID; 25/PK
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX30800-04
Your Organization's Price:
Gemakkelijk overstappen naar een luer-fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: Female luer to 3/16" hose barb
Deze luers zijn gemaakt van dierproefvrij natuurlijk polypropyleen en voldoen aan de Klasse VI-specificaties. Luers bieden superieure chemische weerstand en zijn geschikt voor ETO- en gammasterilisatie.