Masterflex® Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 10-32 UNF; 25/PK
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX45501-60
Your Organization's Price:
Gemakkelijke overgang tussen luer-fittingen en UNF- of NPT-schroefdraad.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 10-32 UNF
Deze adapters passen tussen luer-fittingen en UNF- of NPT-schroefdraad. Er zijn zowel mannelijke als vrouwelijke luer-aansluitingen beschikbaar. Alle schroefdraadverbindingen zijn binnendraad.