Masterflex® Syringe Pump, Push/Pull, Programable, Touchscreen Control; 100/240 VAC
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX74905-58
Your Organization's Price:
Control flow rate via the user-friendly touch-screen display.
  • Flow rates from 5 pL/min to 220 ml/min
  • Full touch-screen enables you to quickly create configurations and recall them later
  • Pump can be placed on its side to provide a smaller footprint with the touch-screen display automatically rotating
  • Accuracy of ±0.3%
These Masterflex® touch-screen syringe pumps offer user-friendly programming and control. All pumps feature an accuracy of ±0.355% and a reproducibility of ±0.05% along with output options RS-232, RS-485, USB, and 15 pin D-sub connector.
<B>Model 74905-58</B> is a programmable version of the 74905-56 pump. It is programmed to accept up to 800 user-defined steps — up to 40 grams with 20 steps each can be stored into memory.
<b>Leveringsinformatie: </b>Includes 6-ft (1.8 m) US power cord.