Masterflex® Extension Cable for Open-Head Sensor Interface; 6 ft (1.8 m) L
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX77097-20
Your Organization's Price:
Make the most of your Masterflex® Open-Head Sensor with these accessories and replacement parts.
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that accept the Easy-Load® II pump head.<br /><br />Use only with I/P® Easy-Load® pump head 77602-30 and with L/S® Easy-Load® II pump heads 77200-30 and 77200-32.