Masterflex® Transfer Tubing, Tygon® E-Lab (E-3603), 3/16" ID x 9/16" OD; 10 Ft
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX06414-10
Your Organization's Price:
Won't crack or oxidize.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 3/16" ID x 9/16" OD
<B>Formulation:</B> Tygon® E-3603 vacuum<br /><B>Applications:</B> General lab, food and beverage, and biopharm. Vacuum service of 29.9” Hg (760 mm Hg) at 73 °F (23 °C).<br /><B>Characteristics:</B> Non-DEHP, phthalate-free plasticizer. Flexible, crack-resistant, nontoxic, and nonoxidizing. Clear.<br /><B>Certifications:</B> USP Class VI and NSF 51<br /><B>Temperature range:</B> Brittle temperature is -51F (-46 °C).<br /><B>Sterilize:</B> Ethylene oxide or autoclave for 30 min at 250 °F (121 °C), 15 psi (1 bar).