Guzzler 44 Hose for model 07090-10 (Tube length: 192)
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX06309-10
Your Organization's Price:
Optimize performance of your Guzzler® Diaphragm Hand Pump.
  • Suitable for use with Guzzler® Portable Diaphragm Hand Pumps
These hoses are noncollapsing, wire-reinforced, and constructed of durable vinyl. <br />Model 06309-10 hose is compatible with Guzzler pumps 07090-09 and -10.<br />Model 06309-11 hose is compatible with Guzzler pump 07090-11.<br />Model 06309-12 hose is compatible with Guzzler pump 07090-12.