Micropump Variable speed drive for low-flow pump heads, 6000 rpm, 0 to 12 VDC
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX07002-37
Your Organization's Price:
Ideal for use with low flow pump heads.
  • Compatible with Micropump® low-flow C-mount pump heads without canisters (order separately)
  • Magnetic seal minimizes friction—all horsepower is transmitted directly to the motor gears
  • Hundreds of hours of dependable, leakproof service
  • Variable speed operation with optional power supply/speed controller—order at side.
Order drives at side; order power supply/speed controller separately at side. Power supply includes a 2-amp fuse and a 6-ft, UL-listed, three-wire cord with plug for 115 VAC operation and measure 7 3/4"L x 5 1/2"W x 4 3/16"H.
MICROPUMP® Micropump Inc., a division of Idex Corp.<br />PTFE® E.I. Du Pont de Nemours & Co.<br />RYTON® Phillips Petroleum Co.