Hand-operated chemical siphon/drum pump, 26 strokes/gallon
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX70607-10
Your Organization's Price:
Ideal for most acid/base or ionic salt applications.
  • Portable and self-priming
  • Suitable for acid and alkali transfer
These plastic hand-operated drum pumps are ideal for water or chemical siphons. All pumps are constructed with PVC barrel and hose. Water siphon model 70607-00 includes a 60” PVC outlet hose that can be cut to fit your application. Chemical siphon models 70607-10 and 70607-12 are suitable for acid and alkali transfer.
Includes PVC barrel and hose, and LDPE tube