Plug Valve, PFA, 1/8" OD compression fittings, 40 psi max
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX98402-05
Your Organization's Price:
Chemically-inert PFA handles most aggressive chemicals.
  • Constructed of durable PFA
Plug valves are FDA-approved and feature PFA seals and nuts.