Masterflex® Stepping-Motor Proportioning Valve, SS; 1/2" OD, 1.5-10 VDC
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR Item#MFLX98651-06
Your Organization's Price:
Resist pressures up to 500 psi!
  • High precision, two-way metering valves in aluminum or 316 SS for air/water
  • Operate continuously without coil overheating problems found in traditional solenoid designs
<B>Stainless steel</B> models include a 316 SS valve blocks, PTFE-lined aluminum housing blocks, Viton® O-rings, and PFA closing pins. <br /><B>Aluminum</B> models include aluminum housings and valve blocks, Viton® O-rings, and PFA closing pins.<br /><br />Also includes a 9-pin "D" electrical connector. Optional connection cable (98651-50) for easy input/output wiring. Power input is +12 VDC at 800 mA, protected by a resettable fuse.