Cell culture plates for adherent cells
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
83.1836 83.1838 83.1839
SARS83.1836EA 0 EUR
SARS83.1836 SARS83.1838 SARS83.1839
Cell culture plates for adherent cells
Microplaten Celcultuurplaten
PS, with lid.
  • Wells with raised rims
  • Stackable
  • Lid with condensation rings can only be placed onto plate in one direction