Filter Holders, Stainless Steel
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
XX3001240 XF2004710 XF2004725
MILFXX3001240EA 728 EUR
MILFXX3001240 MILFXF2004710 MILFXF2004725
Filter Holders, Stainless Steel
Filterhouders
Stainless steel filter holders used to vacuum filter liquids for bacteriological or particulate contamination analysis.
Outlet: No. 8 perforated silicone stopper mounts in standard one liter filtering flask.