Biohazard Bags
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86.1197 86.1198 86.1199 86.1201 86.1202 86.1203 86.1203.103 86.1204 86.1200
SARS86.1197EA 0 EUR
SARS86.1197 SARS86.1198 SARS86.1199 SARS86.1201 SARS86.1202 SARS86.1203 SARS86.1203.103 SARS86.1204 SARS86.1200
Biohazard Bags
Zakken Afvalzakken
These disposal bags are ideal for the containment and disposal of contaminated laboratory and hospital consumables. A 50 µm thick polypropylene bag with a durable wide base seam ensures a good resistance to tear and puncturing. However, in view of the inherent risk of injury, sharp or pointed objects must never be placed into the bags.
  • Durable foil material (50 µm) for improved safety
  • Reduced waste volume
  • Suitable for hot steam sterilisation in the autoclave from 121 °C to approx. 134 °C
Opgelet: For complete sterilisation the disposal bags must always be open during autoclaving.