Ferrules for gas chromatography
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
706246 708308 708353 708354 708355
MANA706246EA 152 EUR
MANA706246 MANA708308 MANA708353 MANA708354 MANA708355
Ferrules for gas chromatography
Fittingen en connectoren
Ferrules for the connection of capillary GC columns.
  • Graphite ferrules provide a temperature stability up to 450 °C
  • Vespel ferrules with 40% graphite have a temperature stability up to 400 °C