Cavity slides for immunofluorescence
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42424010 42423010 42420010 42422010 42421010
631-9485EA 83.4 EUR
631-9485 HECH42423010 HECH42420010 HECH42422010 HECH42421010
Cavity slides for immunofluorescence
Object- en dekglaasjes Objectglaasjes met holte
Black (or blue) epoxy-coated slides.
Specifically designed for immunofluorescence.