Tubing, silicone, EXACANAL
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
760350 760180
228-1607EA 156 EUR
228-1607 228-1194
Tubing, silicone, EXACANAL
Slangen Transfer Tubing
Silicone COVOSIL 9161, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Very flexible
Temperature resistance: –60 to +200 °C
Hardness (Shore durometer A): 60
Density: 1,14
Certificaten: Complies with FDA (21 CFR § 177.2600) standards