Capillary columns, CP-SilicaPLOT
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP8564 CP8568 CP8570 CP8571 CP8567 CP8566 CP8570I5
VARICP8564EA 0 EUR
VARICP8564 VARICP8568 VARICP8570 VARICP8571 VARICP8567 VARICP8566 VARICP8570I5
Capillary columns, CP-SilicaPLOT
Kolommen voor chromatografie
CP-SilicaPLOT brings the benefits of capillary PLOT columns (higher efficiency and faster analysis time) to many applications that previously could only be done by packed columns.
  • No influence of water on retention times for robust methodology
  • Elution of CO₂ and sulfur gases at ppm levels for improved productivity
  • Separates cyclopropane from propylene for accurate results
  • Ideal for CO₂ in ethylene, freons/CFCs, hydrocarbons, propylene and sulfur gases
  • Elutes CO₂ and sulfur gases at ppm levels and separates cyclopropane from propylene
  • Decomposition of