Affinity chromatography columns, TSKgel®
Leverancier: TOSOH Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0008645 0008647 0013066 0014440 0014449 0014451 0014455 0014456 0020022 0013125
TOYO0008645EA 1880 EUR
TOYO0008645 TOYO0008647 TOYO0013066 TOYO0014440 TOYO0014449 TOYO0014451 TOYO0014455 TOYO0014456 TOYO0020022 TOYO0013125
Affinity chromatography columns, TSKgel®
Kolommen voor chromatografie
High efficiency resin based affinity chromatography columns will separate or purify many enzymes and other proteins with high level of specificity and selectivity.
  • Rigid polymeric base resin with high size exclusion limit (>5×10⁶ Da) and small particle size
  • Stable affinity ligands that enhances the access of large proteins to affinity ligands
  • High efficiency for analytical (10 µm) and semi-preparative (13 µm) affinity applications
  • Wide pH range (2 to 12) of the base resin, enabling robust cleaning options
  • Long lifetime, solvent compatibility,