Carboys, narrow neck
Leverancier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B00176990 B00176987 B00176988 B00176991 B00177013 B00176992 B00176993 B00176994 B00176995 B00176997 B00176998 B00176999 B00177000
216-0007EA 148 EUR
216-0007 216-0001 216-0002 216-0004 BEHRB00177013 216-0005 216-0006 718-0002 718-0001 718-0003 718-0006 718-0012 216-8751
Carboys, narrow neck
Flessen Voorraadflessen
These canisters are made from PE.
  • Versatile and resistant to many substances
  • Maximum temperature: 50 to 80 °C
Canisters with welded stopcock keeps the behroplast® canisters shut, even during prolonged heavy use. The canisters can be used wherever liquids are bottled, handled and stored such as food processing and testing, electro-plating technology, general laboratory use, etc.