Tubing kit for aromatic organic solvents
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9910124600 9910124700 9910124800 9910124900
VARI9910124600EA 0 EUR
VARI9910124600 VARI9910124700 VARI9910124800 VARI9910124900
Tubing kit for aromatic organic solvents
Slangen Chromatography Tubing
The ICP-OES tubing kit for aromatic organic solvents includes:
  • Two packs viton peristaltic pump tubing, 12/pack (one each black/black and white/white)
  • One FEP transfer tube (spray chamber to torch)
  • Drain tubing, 1 m
  • Tubing for waste line from spray chamber, 1 m
  • OnePack EzyFit sample inlet connectors, 10/pack
  • Nebulizer capillary tubing, 2 m
  • Barb connectors for joining tubing (5 each 1/16 to 1/8 in Int.Ø. and 5 each 1/16 to 1/16 in Int.Ø.)