PIG® spill caddy
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RFL344-999 RFL244-999
121-0517EA 158 EUR
121-0517 121-0112
PIG® spill caddy
Absorptiematerialen en bescherming tegen morsen
The mobile PIG® spill caddy for fast spill response or daily leak maintenance.
  • High-visibility yellow container is easy to spot when you need it
  • Contents are compartmentalised for easy access
  • Adjustable shelves
  • 20 cm wheels for easy mobility
  • Double-wall polyethylene cart for durability
Container can be locked with a padlock (not included) to prevent pilferage.
Opgelet: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.