Support clips
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
580 582 583 584 581 590 591 592 593 594 595 596 597 598 599 600
BOCH580EA 42.8 EUR
BOCH580 BOCH582 BOCH583 BOCH584 BOCH581 BOCH590 BOCH591 BOCH592 BOCH593 BOCH594 BOCH595 BOCH596 BOCH597 BOCH598 BOCH599 BOCH600
Support clips
Statiefringen
These support clips are coated with ceramic inside.
  • For tubes and cylindrical vessels
  • With fitting
Bestelinformatie: The shafts (rods) need to be ordered separately.