Jars, tall
Leverancier: MURRAY S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G805/03 G805/05 G805/07 G805/A2 G805/A3 G805/A5 G805/A7 G805/A8 G805/08 G805/02
MURRG805/03EA 85.1 EUR
MURRG805/03 MURRG805/05 MURRG805/07 MURRG805/A2 MURRG805/A3 MURRG805/A5 MURRG805/A7 MURRG805/A8 MURRG805/08 MURRG805/02
Jars, tall
Potjes Potjes voor algemeen gebruik
Clear or amber glass, without cap.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Amber coloured to protect light-sensitive materials
Perfect for sample collection, storing and transporting of powders, granulates, pastes or liquids.