Automatic sealer for adhesive tape seals and cap mats, SuperSealer
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
479-1248EA 6675 EUR
479-1248
Automatic sealer for adhesive tape seals and cap mats, SuperSealer
Filmapplicators Afdichtfilmapplicators
Thermo Scientific™ SuperSealer ensures a proper seal on plates, blocks and storage tube racks.
  • Simplifies the sealing of plates, blocks and tube racks of virtually any height, with no adjustments needed, ensuring a secure and consistent seal
  • Once the seal along with the plate, block or tube rack are placed into the drawer, simply close the door to initiate sealing
  • Designed to work with Thermo Scientific Matrix SepraSeal mats and CapMats
Leveringsinformatie: Includes automatic cap, tape and SepraSeal mat applicator.