Conductivity cells, Orion™ 2002SS and 2002CC
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2002CC 2002SS
ORIO2002CCEA 1820 EUR
ORIO2002CC ORIO2002SS
Conductivity cells, Orion™ 2002SS and 2002CC
Elektroden Geleidbaarheidselektroden
These durable conductivity cells provide high performance conductivity measurements.
2002CC:
Sensor features a low cell constant value and is designed to provide high accuracy in low conductivity, ultra pure water and pure water samples in a process environment
Built with 316 stainless steel
Provides months of accurate measurements with virutally no maintenance between calibrations
2002SS:
4-electrode conductivity cell with an epoxy body
Ideal for high and standard conuctivity samples across a variety of industrial applications
Four-electrode design compensates for electrode fouling, cableMore Product Information