Rekken voor Revco UxF en HERAfreeze® HFU T-vriezers
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1950519 1950518 920099
HERA1950519EA 253 EUR
HERA1950519 HERA1950518 HERA920099
Rekken voor Revco UxF en HERAfreeze® HFU T-vriezers
Rekken Rekken voor cryogene dozen
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for Revco™ UxF and HERAfreeze™ HFU T Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deep well formats.