Serial extraction units, R 106 S/R 106 S-SK, behrotest®
Leverancier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B00218424 B00520234
BEHRB00218424EA 5880 EUR
BEHRB00218424 BEHRB00520234
Serial extraction units, R 106 S/R 106 S-SK, behrotest®
Extractors Soxhlet-extractors en Soxhlet-type extractors
Behr equipment for Soxhlet extraction comply with the most various requirements of daily laboratory routines.
  • Practical holders for condenser and extraction adapter for secure placement between extractions
  • Extractor sizes from 30 ml to 1000 ml
  • Condenser with screw connections
Complete compact extraction system with basic rack, heating device, holder, hoses and glassware (reaction flasks, extractor, Dimroth condenser for extraction). Infinitely variable heating regulation After the extraction cycle, the compact system removes the solvent, by means of aMore Product Information