PIG® spill response bucket kit
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
64CRSC KIT213
NEWP64CRSCEA 137 EUR
NEWP64CRSC 121-0556
PIG® spill response bucket kit
Absorptiematerialen en bescherming tegen morsen
Just grab and go for fast response to spills up to 13,8 litres.
  • Container is UN-rated for shipping waste (UN1H2/Y30/S)
  • PIG socks stop spreading spills; PIG mat absorbs quickly
  • Prepacked container provides easy access to contents
  • Bold Spill Kit lettering for quick identification
  • Temporary disposal bags with ties help make cleanup easier
Tamperproof seals help prevent pilfering of spill response supplies.
Opgelet: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.