Digestion system, behrotest®
Leverancier: BEHR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B00218448 B00218430 B00218446
BEHRB00218448EA 3250 EUR
BEHRB00218448 BEHRB00218430 BEHRB00218446
Digestion system, behrotest®
Destructiesystemen Mineralisatiewerkstations
Digestion apparatus for crude fibre or hydrolysis.
Leveringsinformatie: Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.