Bath fluids
Leverancier: Huber
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HUBE6672EA 405 EUR
HUBE6672 HUBE6276 HUBE6155 HUBE6166 HUBE6165 HUBE6275 HUBE6164 HUBE6163 HUBE6258 HUBE6158 HUBE6168 HUBE6157 HUBE6673 HUBE6167 HUBE6156 HUBE9685 HUBE6162 HUBE6161 HUBE10656 HUBE6172 HUBE6171 HUBE6170 HUBE6479 HUBE6259
Bath fluids
Vloeistoffen voor circulatiebaden
Thermal fluids with a wide range of operating temperatures and good thrmodynamic properties for use in circulators and heating systems where water is not appropriate.
All viscosities values are dynamic visosities measured at 25 °C except DW-Therm® which was measured at 20 °C.
Opgelet: * –20...+235 °C for closed system applications
Use biocides safely. Always read the label and product information before use