PIG® high-visibility yellow mat pads and rolls
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAT604 MAT630 MAT605
NEWPMAT604EA 136 EUR
NEWPMAT604 NEWPMAT630 NEWPMAT605
PIG® high-visibility yellow mat pads and rolls
Absorptiematerialen en bescherming tegen morsen
Hi-viz spill absorbents alert workers to slip hazards while cleaning up leaks.
  • High-visibility yellow colour is easier to see in poorly lit areas to minimise tripping hazards; alerts workers to potential dangers and work in progress
  • Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind